Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals
Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals
Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 163670010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS124-40-3
IUPAC Namedimethylamine
Molecular FormulaC2H7N
InChI KeyROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILESCNC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Titration with HCl39.0 to 41.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only