2-Quinoxaloyl chloride, 95%
2-Quinoxaloyl chloride, 95%
2-Quinoxaloyl chloride, 95%
Thermo Scientific Chemicals

2-Quinoxaloyl chloride, 95%

CAS: 54745-92-5 | C9H5ClN2O | 192.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 163762500
Price (TWD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS54745-92-5
IUPAC Namequinoxaline-2-carbonyl chloride
Molecular FormulaC9H5ClN2O
InChI KeySOPDQKNXOCUBSR-UHFFFAOYSA-N
SMILESClC(=O)C1=CN=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Infrared spectrumConforms
GC>=94.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only