N,N'-Methylenebisacrylamide, 96%, extra pure
N,N'-Methylenebisacrylamide, 96%, extra pure
N,N'-Methylenebisacrylamide, 96%, extra pure
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 96%, extra pure

CAS: 110-26-9 | C7H10N2O2 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 163801000
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.00
Save 5.35
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.00
Save 5.35
Each
Chemical Identifiers
CAS110-26-9
IUPAC NameN-[(prop-2-enamido)methyl]prop-2-enamide
Molecular FormulaC7H10N2O2
InChI KeyZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILESC=CC(=O)NCNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
HPLC>=95.0 %
Infrared spectrumConforms
Water=<1 % (K.F.)
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only