1-Ethyl-2-pyrrolidinone, 98%
1-Ethyl-2-pyrrolidinone, 98%
1-Ethyl-2-pyrrolidinone, 98%
Thermo Scientific Chemicals

1-Ethyl-2-pyrrolidinone, 98%

CAS: 2687-91-4 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 163931000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2687-91-4
IUPAC Name1-ethylpyrrolidin-2-one
Molecular FormulaC6H11NO
InChI KeyZFPGARUNNKGOBB-UHFFFAOYSA-N
SMILESCCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4640 to 1.4680 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only