2-Methyl-4-nitroaniline, 99%
2-Methyl-4-nitroaniline, 99%
2-Methyl-4-nitroaniline, 99%
Thermo Scientific Chemicals

2-Methyl-4-nitroaniline, 99%

CAS: 99-52-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Catalog NumberQuantity
1639500505 g
Catalog number 163950050
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS99-52-5
IUPAC Name2-methyl-4-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyXTTIQGSLJBWVIV-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
Appearance (Color)Yellow to brown to khaki
GC>=98.5 %
Infrared spectrumConforms
Melting point129°C to 133°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only