7-Amino-1,3-naphthalenedisulfonic acid, Tech.
7-Amino-1,3-naphthalenedisulfonic acid, Tech.
7-Amino-1,3-naphthalenedisulfonic acid, Tech.
Thermo Scientific Chemicals

7-Amino-1,3-naphthalenedisulfonic acid, Tech.

CAS: 86-65-7 | C10H9NO6S2 | 303.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 163965000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS86-65-7
IUPAC Name7-aminonaphthalene-1,3-disulfonic acid
Molecular FormulaC10H9NO6S2
InChI KeyCMOLPZZVECHXKN-UHFFFAOYSA-N
SMILESNC1=CC=C2C=C(C=C(C2=C1)S(O)(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to green-beige to light brown
Infrared spectrumConforms
Titration with NaOH>=60 % (As is)
Appearance (Form)Powder or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only