Heptanoic acid, 98%
Heptanoic acid, 98%
Heptanoic acid, 98%
Thermo Scientific Chemicals

Heptanoic acid, 98%

CAS: 111-14-8 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 164170025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-14-8
IUPAC Nameheptanoic acid
Molecular FormulaC7H14O2
InChI KeyMNWFXJYAOYHMED-UHFFFAOYSA-N
SMILESCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Titration with NaOH>=97.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4210 to 1.4250 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only