Benzyltriethylammonium bromide, 99%
Benzyltriethylammonium bromide, 99%
Benzyltriethylammonium bromide, 99%
Thermo Scientific Chemicals

Benzyltriethylammonium bromide, 99%

CAS: 5197-95-5 | C13H22BrN | 272.23 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 164201000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5197-95-5
Appearance (Form)Crystalline powder
Melting point187°C to 197°C (Dec)
Appearance (Color)White to cream to light yellow
Infrared spectrumConforms
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only