4-Amino-2-nitrophenol, 99%
4-Amino-2-nitrophenol, 99%
4-Amino-2-nitrophenol, 99%
Thermo Scientific Chemicals

4-Amino-2-nitrophenol, 99%

CAS: 119-34-6 | C6H6N2O3 | 154.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1642300505 gGlass bottle
Catalog number 164230050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS119-34-6
IUPAC Name4-amino-2-nitrophenol
Molecular FormulaC6H6N2O3
InChI KeyWHODQVWERNSQEO-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point124°C to 128°C
GC>=98.5 %
Appearance (Color)Dark brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only