Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
Have Questions?
Change view
Catalog Number
Tray Layers
Culture Area
No. per Case
164327
10
6320 cm²
2/Cs.
139446
40
25,280 cm²
2/Cs.
140004TS
also known as 140004
4
2528 cm²
10/Cs.
165250
1
632 cm²
8/Cs.
167695
2
1264 cm²
6/Cs.
170009
10
6320 cm²
6/Cs.
6 Options
Catalog number 164327
Price (USD)
738.00
Case of 2
Tray Layers:
10
Culture Area:
6320 cm²
No. per Case:
2/Cs.
Price (USD)
738.00
Case of 2
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
Ideal for adherent cells
Boost production efficiency with more surface area in a small footprint
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure TypeAdapter Cap and Port Cover Cap
DescriptionOriginal Cell Factory
Green FeaturesISCC PLUS
No. per Case2/Cs.
No. per Pack1 Ea.
Product TypeCell Factory System
Surface CoatingNunclon™ Delta
Tray Layers10
Bottom ShapeFlat
Culture Area6320 cm²
MaterialPolystyrene
Surface TreatmentNunclon Delta
Unit SizeCase of 2
Frequently asked questions (FAQs)
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?
Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.
The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells
What volume of medium should I use for Nunc Cell Factory Systems?
We recommend a 200 mL volume of medium per tray (632 cm^2).
Therefore the minimum volumes we recommend are as follows:
Cat. No.; No. per Pack/No. per Case; No. of Trays; Minimum Volume (mL)
- 165250; 1/8; 1-Tray; 200 mL
- 167695; 1/5; 2-Tray; 400 mL
- 164327; 1/2; 10-Tray; 2000 mL
- 170009; 1/6; 10-Tray; 2000 mL
- 139446; 1/2; 40-Tray; 8000 mL
What are the dimensions and culture areas for each tray version of your Nunc Cell Factory Systems?
Please review the following information regarding the dimension and culture areas for each tray version of the Nunc Cell Factory Systems:
Cat. No.; No. per Pack/No. per Case; No. of Tray Layers; Dimensions L x W x H (mm); Culture Area (cm^2)
- 165250; 1/8; 1-Tray; 335 mm x 205 mm x 37 mm; 632 cm^2
- 167695; 1/6; 2-Tray; 335 mm x 205 mm x 52 mm; 1264 cm^2
- 164327; 1/2; 10-Tray; 335 mm x 205 mm x 190 mm; 6320 cm^2
- 170009; 1/6; 10-Tray; 335 mm x 205 mm x 190 mm; 6320 cm^2
- 139446; 1/2; 40-Tray; 335 mm x 205 mm x 700 mm; 25284 cm^2
Can I view cells in a 10-tray Nunc Cell Factory System under a microscope?
Due to the height of the 10-tray and 40-tray Nunc Cell Factory Systems, cells cannot be viewed with common microscopes. Typically cells are seeded in a 1-tray or 2-tray cell factory system as a control. Cells grown in each of these products can be viewed under a microscope.
Alternatively, inverted stereo microscopes, with a powerful light source on the view side, and adjusted for the height of the cell factory system have been used to view the first few layers.
What are your Nunc Cell Factory Systems?
Our Nunc Cell Factory Systems are designed for large scale cell culture and production of biomaterials such as vaccines, monoclonal antibodies, etc. They provide large growth surface in limited space areas and have the Nunclon Delta surface coating.
Some benefits are the low risk of contamination and the easy to handle multi-layer stacks. Nunc Cell Factory Systems are available with 1-, 2-, 10-, and 40-tray versions.