Pregnenolone, 99%, Thermo Scientific Chemicals
Pregnenolone, 99%, Thermo Scientific Chemicals
Pregnenolone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pregnenolone, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量パッケージ
16442025025gGlass bottle
1644200505gGlass bottle
164421000100gGlass bottle
製品番号(カタログ番号) 164420250
価格(JPY)
-
数量:
25g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS145-13-1
IUPAC Name1-[(1S,3aS,3bS,7S,9aR,9bS,11aS)-7-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]ethan-1-one
Molecular FormulaC21H32O2
InChI KeyORNBQBCIOKFOEO-QGVNFLHTSA-N
SMILESCC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
さらに表示
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
Specific optical rotation+27° to +30° (20°C, 589 nm) (c=1, EtOH)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only