Betaine hydrochloride, 99%, extra pure, specified according to the requirements of USP
Betaine hydrochloride, 99%, extra pure, specified according to the requirements of USP
Betaine hydrochloride, 99%, extra pure, specified according to the requirements of USP
Thermo Scientific Chemicals

Betaine hydrochloride, 99%, extra pure, specified according to the requirements of USP

CAS: 590-46-5 | C5H12ClNO2 | 153.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 164550025
Price (USD)
276.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
276.00
Each
Chemical Identifiers
CAS590-46-5
IUPAC Name(carboxymethyl)trimethylazanium chloride
Molecular FormulaC5H12ClNO2
InChI KeyHOPSCVCBEOCPJZ-UHFFFAOYSA-N
SMILES[Cl-].C[N+](C)(C)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Identification BPasses test
Water=<0.5 %
pH0.8 to 1.2 (1 in 4 solution)
Infrared spectrumConforms (identification A)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only