DL-Phenylalanine, 99%
DL-Phenylalanine, 99%
DL-Phenylalanine, 99%
Thermo Scientific Chemicals

DL-Phenylalanine, 99%

DL-Phenylalanine, 99%, C9H11NO2, CAS Number-150-30-1, 2-amino-3-phenylpropionic acid, alanine, phenyl-, dl, dl-2-amino-3-phenylpropanoic acid, dl-3-phenylalanine, dlpa, dl-phenylalanine, h-dl-phe-oh, phenylalanin, phenylalanine, phenylalanine, dl, 100g, 14,498, CHEBI:28044, White, 205-756-7, 165.19 | CAS: 150-30-1 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 164570250
Price (EUR)
27,65
Online exclusive
30,40
Save 2,75 (9%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
27,65
Online exclusive
30,40
Save 2,75 (9%)
Each
Chemical Identifiers
CAS150-30-1
IUPAC Name2-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyCOLNVLDHVKWLRT-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.3 %
Arsenic (As)=<3 ppm
Appearance (Form)Powder or crystals
Titration with HClO498.5 to 101.5 % (on dry substance)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only