Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom, Optical Coverglass Base Microplate
Thermo Scientific™

Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom, Optical Coverglass Base Microplate

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Coverglass Base
Have Questions?
카탈로그 번호수량
164588Case of 30
카탈로그 번호 164588
제품 가격(KRW)
2,259,000
온라인 행사
Ends: 31-Dec-2026
2,657,000
할인액 398,000
Case of 30
수량:
Case of 30
제품 가격(KRW)
2,259,000
온라인 행사
Ends: 31-Dec-2026
2,657,000
할인액 398,000
Case of 30
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.

Features:

  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Cell culture surface is ideal for cell-based assays
  • CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
  • Customized bar coding available
사양
인증/적합성All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
뚜껑With Lid
케이스당 수30
팩당 수6
포장6 Pk., 30 Cases
제품 유형Microplate
수량Case of 30
어레이8 x 12
색상Black, Clear Bottom
물질Polystyrene
웰 수96
제품라인Nunc
멸균Sterile
표면 처리Cell Culture
웰 형상Flat Bottom (F-well)
Unit SizeCase of 30

자주 묻는 질문(FAQ)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

How does cell culturing on Nunc 384-Well Microplates that have a cell culture-treated surface compare with 96-Well Microplates that have a cell culture-treated surface?

The cell culture-treated surfaces are very similar but the wells of the Nunc 384-Well Microplates are smaller than the wells of the Nunc 96-Well Microplate.
Culture recommendations for the Nunc 384-Well Microplates are:
- 15 - 50 µL media per well
- 0.5 - 2.0 x 10^4 cells per well

Should I use an opaque black/white immuno plate, or an "optical-bottom" design?

Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.