Adenosine 5'-diphosphate, disodium salt hydrate, 98%
Adenosine 5'-diphosphate, disodium salt hydrate, 98%
Adenosine 5'-diphosphate, disodium salt hydrate, 98%
Thermo Scientific Chemicals

Adenosine 5'-diphosphate, disodium salt hydrate, 98%

CAS: 16178-48-6 | C10H13N5Na2O10P2 | 471.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1646700101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 164670010
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Chemical Identifiers
CAS16178-48-6
IUPAC Namedisodium hydrogen ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC10H13N5Na2O10P2
InChI KeyORKSTPSQHZNDSC-WCYUCLFNNA-L
SMILES[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Ratio of Absorbances0.76 to 0.80 (A250/A260)
Appearance (Color)White
Infrared spectrumConforms
UV spectrophotometry>=82.0 % (ADP, on as is basis) (lambda max. 260 nm)
Sodium (Na)8 to 10 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only