4-Mercaptopyridine, 96%
4-Mercaptopyridine, 96%
4-Mercaptopyridine, 96%
Thermo Scientific Chemicals

4-Mercaptopyridine, 96%

CAS: 4556-23-4 | C5H5NS | 111.16 g/mol
数量:
1 g
10 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 164710010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4556-23-4
IUPAC Name1,4-dihydropyridine-4-thione
Molecular FormulaC5H5NS
InChI KeyFHTDDANQIMVWKZ-UHFFFAOYSA-N
SMILESS=C1C=CNC=C1
さらに表示
Appearance (Form)Crystalline powder
Titration with NaOH>=95 %
Appearance (Color)Yellow to yellow-green to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only