Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™
Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™
Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™
Thermo Scientific Chemicals

Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™

CAS: 41233-93-6 | C5H11KO | 126.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 164728000
Price (EUR)
253,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
253,00
Each
Chemical Identifiers
CAS41233-93-6
IUPAC Namepotassium 2-methylbutan-2-olate
Molecular FormulaC5H11KO
InChI KeyZRLVQFQTCMUIRM-UHFFFAOYSA-N
SMILES[K+].CCC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Titration with HClO40.85 to 1.10 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only