Find Nucleic Acid Purification and Analysis technical tips, troubleshooting help, and resources within our new Nucleic Acid Purification and Analysis Support Center.
UltraPure™ Agarose
Invitrogen™

UltraPure™ Agarose

UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
16500500500 g
16500100100 g
Catalog number 16500500
Price (USD)
824.65
Online exclusive
896.00
Save 71.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
824.65
Online exclusive
896.00
Save 71.35
Each
UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose is ideal for resolving DNA and RNA fragments from 100 bp to >30 kb. Features of UltraPure™ Agarose:

• Ideal for analysis and recovery of DNA and RNA for routine applications
• Strong gel structure allows for better handling and less breakage
• Can be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)

Improved packaging
The new environmentally friendly packaging uses 75% less plastic than the original bottles. This means less energy and raw material used in manufacture, and less waste in landfills. Additionally, the easy-pour spout reduces the likelihood of spills and contamination.

Performance and quality testing
The performance of UltraPure™ Agarose is evaluated to satisfy set specifications in appearance, moisture, gel strength, gelling temperature, melting temperature, sulfate content, electroendosmosis, and DNase/RNase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Nucleic Acid Gel Electrophoresis, Protein Electrophoresis, Blotting
FormPowder
Gel CompatibilityAgarose Gels
Melting PointStandard Melting Point
No. of Reactions2500
Quantity500 g
Shipping ConditionRoom Temperature
Product LineUltraPure
Product TypeAgarose Gel
Separation Range100 to >30000 bp
Unit SizeEach
Contents & Storage
Contents: 1 pouch containing 500 g of UltraPure™ Agarose
Storage: 15°C to 30°C

Frequently asked questions (FAQs)

I want to pour my own gels. Which agarose should I use?

Our UltraPure Agarose is standard melting-point agarose designed for routine separation and analysis of DNA and RNA fragments in the 500-23,000 nt range. UltraPure Agarose 1000 is a specialized agarose that provides higher resolution of PCR fragments and other short DNA fragments. We also offer an UltraPure Low Melting Point Agarose, which is ideal for resolving DNA fragments from 10 to 1,000 bp with a low melting temperature of 65°C or less.

How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.

Citations & References (20)

Citations & References
Abstract
Proteomic analysis reveals presence of platelet microparticles in endothelial progenitor cell cultures.
Authors:Prokopi M, Pula G, Mayr U, Devue C, Gallagher J, Xiao Q, Boulanger CM, Westwood N, Urbich C, Willeit J, Steiner M, Breuss J, Xu Q, Kiechl S, Mayr M,
Journal:Blood
PubMed ID:19369228
'The concept of endothelial progenitor cells (EPCs) has attracted considerable interest in cardiovascular research, but despite a decade of research there are still no specific markers for EPCs and results from clinical trials remain controversial. Using liquid chromatography-tandem mass spectrometry, we analyzed the protein composition of microparticles (MPs) originating from ... More
A phantom for diffusion-weighted MRI (DW-MRI).
Authors:Lavdas I, Behan KC, Papadaki A, McRobbie DW, Aboagye EO,
Journal:J Magn Reson Imaging
PubMed ID:23576443
'PURPOSE: To develop tissue-equivalent diffusivity materials and build a spherical diffusion phantom which mimics the conditions typically found in biological tissues. Also, to assess the reproducibility of ADC measurements from a whole-body diffusion protocol. MATERIALS AND METHODS: Nickel-doped agarose/sucrose gels were manufactured and used to build a spherical diffusion phantom ... More
Determinants of nucleosome organization in primary human cells.
Authors:Valouev A, Johnson SM, Boyd SD, Smith CL, Fire AZ, Sidow A,
Journal:Nature
PubMed ID:21602827
'Nucleosomes are the basic packaging units of chromatin, modulating accessibility of regulatory proteins to DNA and thus influencing eukaryotic gene regulation. Elaborate chromatin remodelling mechanisms have evolved that govern nucleosome organization at promoters, regulatory elements, and other functional regions in the genome. Analyses of chromatin landscape have uncovered a variety ... More
Controlling cell position in complex heterotypic 3D microtissues by tissue fusion.
Authors:Rago AP, Dean DM, Morgan JR,
Journal:Biotechnol Bioeng
PubMed ID:19012266
'Tissue fusion and cell sorting are processes fundamental to developmental biology with applications in tissue engineering. We have designed a fusion assay to investigate the factors governing the fusion of microtissues and the cell sorting that occurs after fusion. Normal human fibroblast (NHF) spheroids were self-assembled and cultured for 1, ... More
Host alternation of chikungunya virus increases fitness while restricting population diversity and adaptability to novel selective pressures.
Authors:Coffey LL, Vignuzzi M,
Journal:J Virol
PubMed ID:21047966
'The mechanisms by which RNA arboviruses, including chikungunya virus (CHIKV), evolve and maintain the ability to infect vertebrate and invertebrate hosts are poorly understood. To understand how host specificity shapes arbovirus populations, we studied CHIKV populations passaged alternately between invertebrate and vertebrate cells (invertebrate ? vertebrate) to simulate natural alternation ... More