2,5-Dihydroxybenzoic acid, 99%
2,5-Dihydroxybenzoic acid, 99%
2,5-Dihydroxybenzoic acid, 99%
Thermo Scientific Chemicals

2,5-Dihydroxybenzoic acid, 99%

CAS: 490-79-9 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 165200500
Price (USD)
120.00
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
120.00
Each
Chemical Identifiers
CAS490-79-9
IUPAC Name2,5-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyWXTMDXOMEHJXQO-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point204°C to 207°C
Titration with NaOH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only