L(+)-Glutamic acid hydrochloride, 99%
L(+)-Glutamic acid hydrochloride, 99%
L(+)-Glutamic acid hydrochloride, 99%
Thermo Scientific Chemicals

L(+)-Glutamic acid hydrochloride, 99%

CAS: 138-15-8 | C5H10ClNO4 | 183.588 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 165235000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS138-15-8
IUPAC Namehydrogen 2-aminopentanedioic acid chloride
Molecular FormulaC5H10ClNO4
InChI KeyRPAJSBKBKSSMLJ-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(CCC(O)=O)C(O)=O
さらに表示
Loss on drying=<0.5 % (105°C, 5 hrs)
Sulfated ash=<0.1 %
Ammonium (NH4)=<200 ppm
Iron (Fe)=<10 ppm
pH1 to 2.5 (2.5 % at 25°C)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only