Nunc™ 96-Well Optical-Bottom Microplate, black, TC surface
Nunc™ 96-Well Optical-Bottom Microplate, black, TC surface
Nunc™ 96-Well Optical-Bottom Microplate, black, TC surface
Thermo Scientific™

Nunc™ 96-Well Optical-Bottom Microplate, black, TC surface

Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Nunc MicroWell 384-well optical-bottom plates, featuring a polystyrene black upper structure with a polystyrene film at the bottom of the plate.
Have Questions?
Catalog NumberQuantity
16530530/Cs.
Catalog number 165305
Price (USD)
431.00
Case of 30
Quantity:
30/Cs.
Price (USD)
431.00
Case of 30

Thermo Scientific Nunc MicroWell 96-well optical-bottom plates with polymer base have a solid polystyrene black upper structure with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.

Black polystyrene upper structure with polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.

  • Background, autoflorescence, and crosstalk are improved when you use Nunc black microplates
  • Black microplates are recommended for florescence measurements with minimum back-scattered light and background fluorescence
  • Also available in white; recommended for luminescence measurements with maximum reflection and minimal autoluminescence
  • Polymer base plate
  • Working range: 50–200 μL/well
  • Customized bar coding is available

Color: black with clear bottom

Sterility: sterile

Lid: with lid

Type: optical polymer base

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWith Lid
No. per Case30
No. per Pack10
Packaging10 Pk., 30 Cases
Product TypeOptical Polymer Base
Quantity30/Cs.
Volume (Metric) Well400 μL
Volume (Metric) Working50 to 200 mL
Array8 x 12
ColorBlack, Clear Bottom
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentCell Culture
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 30

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

How does cell culturing on Nunc 384-Well Microplates that have a cell culture-treated surface compare with 96-Well Microplates that have a cell culture-treated surface?

The cell culture-treated surfaces are very similar but the wells of the Nunc 384-Well Microplates are smaller than the wells of the Nunc 96-Well Microplate.
Culture recommendations for the Nunc 384-Well Microplates are:
- 15 - 50 µL media per well
- 0.5 - 2.0 x 10^4 cells per well

Should I use an opaque black/white immuno plate, or an "optical-bottom" design?

Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.