N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%
Thermo Scientific Chemicals

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%

CAS: 16357-59-8 | C14H17NO3 | 247.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 165350250
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
60.60
Save 8.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
60.60
Save 8.95
Each
Chemical Identifiers
CAS16357-59-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=99.0 %
Appearance (Form)Crystalline powder
Appearance (Color)White to beige
Melting point63°C to 66°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only