6-Bromohexanoic acid, 98%
6-Bromohexanoic acid, 98%
6-Bromohexanoic acid, 98%
Thermo Scientific Chemicals

6-Bromohexanoic acid, 98%

CAS: 4224-70-8 | C6H11BrO2 | 195.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 165671000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4224-70-8
IUPAC Name6-bromohexanoate
Molecular FormulaC6H10BrO2
InChI KeyNVRVNSHHLPQGCU-UHFFFAOYSA-M
SMILES[O-]C(=O)CCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystals
Infrared spectrumConforms
Melting point32°C to 35°C
Titration with NaOH>=97.5 %
Appearance (Color)White to light orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only