Decyl alcohol, 98+%
Decyl alcohol, 98+%
Decyl alcohol, 98+%
Thermo Scientific Chemicals

Decyl alcohol, 98+%

CAS: 112-30-1 | C10H22O | 158.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 165760025
Price (USD)
107.65
Online exclusive
119.00
Save 11.35
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
107.65
Online exclusive
119.00
Save 11.35
Each
Chemical Identifiers
CAS112-30-1
IUPAC Namedecan-1-ol
Molecular FormulaC10H22O
InChI KeyMWKFXSUHUHTGQN-UHFFFAOYSA-N
SMILESCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Refractive index1.4350 to 1.4390 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only