Ninhydrin, spectrophotometric grade
Ninhydrin, spectrophotometric grade
Ninhydrin, spectrophotometric grade
Acros Organics

Ninhydrin, spectrophotometric grade

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 165870250
HSN 29144000
Price (INR)
8,592.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,592.00
Each
Chemical Identifiers
CAS485-47-2
IUPAC Name2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
Molecular FormulaC9H6O4
InChI KeyFEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILESOC1(O)C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Suitability(for amino acid analysis) Passes test
pH4.0 to 5.6 (1 % in water)
Sulfated ash=<0.1 %
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only