1-(2-Pyridyl)piperazine, 98%
1-(2-Pyridyl)piperazine, 98%
1-(2-Pyridyl)piperazine, 98%
Thermo Scientific Chemicals

1-(2-Pyridyl)piperazine, 98%

CAS: 34803-66-2 | C9H13N3 | 163.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 165900050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS34803-66-2
IUPAC Name1-(pyridin-2-yl)piperazine
Molecular FormulaC9H13N3
InChI KeyGZRKXKUVVPSREJ-UHFFFAOYSA-N
SMILESC1CN(CCN1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Refractive index1.5930 to 1.5960 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only