Fumaryl chloride, 95%
Fumaryl chloride, 95%
Fumaryl chloride, 95%
Thermo Scientific Chemicals

Fumaryl chloride, 95%

CAS: 627-63-4 | C4H2Cl2O2 | 152.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 165971000
Price (EUR)
129,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
129,00
Each
Chemical Identifiers
CAS627-63-4
IUPAC Name(2E)-but-2-enedioyl dichloride
Molecular FormulaC4H2Cl2O2
InChI KeyZLYYJUJDFKGVKB-OWOJBTEDSA-N
SMILESClC(=O)C=CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=94 %
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.4980 to 1.5010 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only