Methyl oxalyl chloride, 97%
Methyl oxalyl chloride, 97%
Methyl oxalyl chloride, 97%
Thermo Scientific Chemicals

Methyl oxalyl chloride, 97%

CAS: 5781-53-3 | C3H3ClO3 | 122.51 g/mol
Have Questions?
Catalog NumberQuantityPackaging
16600025025 mLGlass bottle
Catalog number 166000250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5781-53-3
IUPAC Namemethyl oxalochloridate
Molecular FormulaC3H3ClO3
InChI KeyZXUQEPZWVQIOJE-UHFFFAOYSA-N
SMILESCOC(=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<200 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4179 to 1.4199 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only