N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
Thermo Scientific Chemicals

N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.

CAS: 7570-45-8 | C15H13NO | 223.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166030250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7570-45-8
IUPAC Name9-ethyl-9H-carbazole-3-carbaldehyde
Molecular FormulaC15H13NO
InChI KeyQGJXVBICNCIWEL-UHFFFAOYSA-N
SMILESCCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or granules
Infrared spectrumConforms
Appearance (Color)Yellow to ochre
Melting point85°C to 91°C
HPLC>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only