Product Image
Thermo Scientific™

CEDIA™ Opiate Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Opiate Drugs of Abuse Assays.
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
16612481 x 495 mL, LC
100098TS1 x 65 mL, MCC
100164293 x 17 mL, Indiko
1000893 x 17 mL, TSC
產品號碼 1661248
價格 (HKD)
-
Quantity:
1 x 495 mL, LC
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Opiate Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
規格
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Opiate Drugs of Abuse Assays
Quantity1 x 495 mL, LC
Sample TypeUrine
TypeReagent
Detectable AnalytesOpiates (OPI)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
內容物與存放
2°C to 8°C