L-Histidine, 98%
L-Histidine, 98%
L-Histidine, 98%
Thermo Scientific Chemicals

L-Histidine, 98%

CAS: 71-00-1 | C6H9N3O2 | 155.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
Catalog number 166155000
Price (USD)
425.65
Online exclusive
473.00
Save 47.35
Each
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
425.65
Online exclusive
473.00
Save 47.35
Each
Chemical Identifiers
CAS71-00-1
IUPAC Name2-amino-3-(1H-imidazol-5-yl)propanoic acid
Molecular FormulaC6H9N3O2
InChI KeyHNDVDQJCIGZPNO-UHFFFAOYNA-N
SMILESNC(CC1=CN=CN1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<500 ppm
Sulfate (SO4)=<300 ppm
Iron (Fe)=<30 ppm
pH7.0 to 8.5 (2 % in water)
Appearance (Color)White to almost white to cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only