L-Methionine, 98+%
L-Methionine, 98+%
L-Methionine, 98+%
Thermo Scientific Chemicals

L-Methionine, 98+%

CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 166165000
Price (USD)
236.00
Each
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
236.00
Each
Chemical Identifiers
CAS63-68-3
IUPAC Name(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular FormulaC5H11NO2S
InChI KeyFFEARJCKVFRZRR-BYPYZUCNSA-N
SMILESCSCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO498.0 to 102.0 % (on dried basis)
Heavy metals (as Pb)=<15 ppm
pH5.5 to 6.5 (1 % in water)
Chloride (Cl)=<500 ppm
Iron (Fe)=<30 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only