Thermo Scientific Chemicals

Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals

Catalog number: 166210500
50 mL, Each
Thermo Scientific Chemicals

Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals

Catalog number: 166210500
50 mL, Each
Quantity

Chemical Identifiers

CAS
1186-52-3
IUPAC Name
(²H₃)ethan(²H)oic acid
Molecular Formula
C2H4O2
InChI Key
QTBSBXVTEAMEQO-GUEYOVJQSA-N
SMILES
[2H]OC(=O)C([2H])([2H])[2H]
Appearance (Color)
Clear colorless
Water
=<0.1 % (Coulometric)
Appearance (Form)
Liquid
D-Enrichment
>=99.4 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text