Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | CEDIA High Calibrator |
Concentration | PPX 5000 ng/mL, Methadone 1000 ng/mL |
Detectable Analytes | PPx, Methadone |
Sample Type | Urine |
Type | Calibrator |
Packaging | 1 Bottle |
Quantity | 1 x 5 mL |
Unit Size | Each |
Description | CEDIA Cutoff Calibrator |
Concentration | PPx 300 ng/mL, Methadone 300 ng/mL |
Detectable Analytes | PPx, Methadone |
Sample Type | Urine |
Type | Calibrator |
Packaging | 1 Bottle |
Quantity | 1 x 5 mL |
Unit Size | Each |
Description | CEDIA Intermediate Calibrator |
Concentration | PPX 1200 ng/mL, Methadone 600 ng/mL |
Detectable Analytes | PPx, Methadone |
Sample Type | Urine |
Type | Calibrator |
Packaging | 1 Bottle |
Quantity | 1 x 5 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Concentration | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
1662864 | Each | PPX 5000 ng/mL, Methadone 1000 ng/mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1662848 | Each | PPx 300 ng/mL, Methadone 300 ng/mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1662856 | Each | PPX 1200 ng/mL, Methadone 600 ng/mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|