Deuterium oxide, for NMR, 100.0 atom % D
Deuterium oxide, for NMR, 100.0 atom % D
Deuterium oxide, for NMR, 100.0 atom % D
Thermo Scientific Chemicals

Deuterium oxide, for NMR, 100.0 atom % D

CAS: 7789-20-0 | H2O | 20.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
100 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166310100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-20-0
IUPAC Name[(²H)oxy](²H)
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES[2H]O[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only