4-Methoxyphenylhydrazine hydrochloride, 98%
4-Methoxyphenylhydrazine hydrochloride, 98%
4-Methoxyphenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

4-Methoxyphenylhydrazine hydrochloride, 98%

CAS: 19501-58-7 | C7H10N2O·ClH | 174.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 166490500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19501-58-7
IUPAC Name(4-methoxyphenyl)hydrazine hydrochloridyl
Molecular FormulaC7H10ClN2O
InChI KeyUNLQTHFXAIISEP-UHFFFAOYSA-N
SMILES[Cl].COC1=CC=C(NN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light pink to grey to purple
Appearance (Form)Powder or crystals and/or chunks
Titration with NaOH97.5 to 102.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only