4-Bromobenzotrifluoride, 99%
4-Bromobenzotrifluoride, 99%
4-Bromobenzotrifluoride, 99%
Thermo Scientific Chemicals

4-Bromobenzotrifluoride, 99%

CAS: 402-43-7 | C7H4BrF3 | 225.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number 166670250
Price (USD)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS402-43-7
IUPAC Name1-bromo-4-(trifluoromethyl)benzene
Molecular FormulaC7H4BrF3
InChI KeyXLQSXGGDTHANLN-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4715 to 1.4745 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only