1,4-Naphthoquinone, 99%, contains up to 6% water
1,4-Naphthoquinone, 99%, contains up to 6% water
1,4-Naphthoquinone, 99%, contains up to 6% water
Thermo Scientific Chemicals

1,4-Naphthoquinone, 99%, contains up to 6% water

CAS: 130-15-4 | C10H6O2 | 158.16 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
產品號碼 166711000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS130-15-4
IUPAC Name1,4-dihydronaphthalene-1,4-dione
Molecular FormulaC10H6O2
InChI KeyFRASJONUBLZVQX-UHFFFAOYSA-N
SMILESO=C1C=CC(=O)C2=CC=CC=C12
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder
Water=<6 % (K.F.)
Infrared spectrumConforms
Appearance (Color)Khaki
GC>=99 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only