N-Acetyl-4-piperidone, 97%
N-Acetyl-4-piperidone, 97%
N-Acetyl-4-piperidone, 97%
Thermo Scientific Chemicals

N-Acetyl-4-piperidone, 97%

CAS: 32161-06-1 | C7H11NO2 | 141.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 166820250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS32161-06-1
IUPAC Name1-acetylpiperidin-4-one
Molecular FormulaC7H11NO2
InChI KeyNNFOVLFUGLWWCL-UHFFFAOYSA-N
SMILESCC(=O)N1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear orange
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5015 to 1.5035 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only