Product Image
Thermo Scientific™

DRI™ Therapeutic Drug Monitoring (TDM) Assays, Digoxin

Receive reliable performance on a wide range of analyzers with Thermo Scientific™ DRI™ Therapeutic Drug Monitoring (TDM) Assays.
Have Questions?
Catalog NumberQuantity
166925 x 8 mL
Catalog number 1669
Price (USD)
-
Quantity:
25 x 8 mL
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ DRI™ Therapeutic Drug Monitoring (TDM) Assays. Thermo Scientific TDM assays have been manufactured since 1986 and provide an excellent range of Thermo Scientific™ CEDIA TDM Assays, Thermo Scientific™ CEDIA TDM Calibrators and DRI TDM Assays with a wide dynamic measuring range of analytes.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDigoxin Assay
Quantity25 x 8 mL
Sample TypePlasma, Serum
TypeReagent
Detectable AnalytesDigoxin
Unit SizeEach
Contents & Storage
2°C to 8°C