4-Methylcyclohexanol, 99%, mixture of cis and trans
4-Methylcyclohexanol, 99%, mixture of cis and trans
4-Methylcyclohexanol, 99%, mixture of cis and trans
Thermo Scientific Chemicals

4-Methylcyclohexanol, 99%, mixture of cis and trans

CAS: 589-91-3 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 166900010
Price (USD)
201.65
Special offer
Online exclusive
Ends: 30-Jun-2026
237.00
Save 35.35 (15%)
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
201.65
Special offer
Online exclusive
Ends: 30-Jun-2026
237.00
Save 35.35 (15%)
Each
Chemical Identifiers
CAS589-91-3
IUPAC Name4-methylcyclohexan-1-ol
Molecular FormulaC7H14O
InChI KeyMQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILESCC1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4570 to 1.4590 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only