Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Thermo Scientific™

Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

Color: Clear; Sterility: Sterile; Lid: With Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsNo. per PackNo. per Case
167008961 Microplate50 Microplates
1610939610 Microplates160 Microplates
Catalog number 167008
Price (USD)
392.65
Online exclusive
418.00
Save 25.35
Case of 50
No. of Wells:
96
No. per Pack:
1 Microplate
No. per Case:
50 Microplates
Price (USD)
392.65
Online exclusive
418.00
Save 25.35
Case of 50

Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001:2008, ISO 13485:2003
CertifiedNon-pyrogenic
Description96F well plate, Nunclon Delta, clear, with lid, Sterile (Pack of 1, Case of 50)
FormatStandard
LidWith Lid
No. per Case50 Microplates
No. per Pack1 Microplate
Packaging1/Pk., 50/Cs.
Product TypeMicroplate
Quantity50/Cs.
Volume (Metric) Well400 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 50

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .