DL-Asparagine monohydrate, 98%
DL-Asparagine monohydrate, 98%
DL-Asparagine monohydrate, 98%
Thermo Scientific Chemicals

DL-Asparagine monohydrate, 98%

CAS: 3130-87-8 | C4H8N2O3 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Catalog number 167120250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS69833-18-7
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Water10.5 to 13.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only