4-tert-Amylphenol, 99%, Thermo Scientific Chemicals
4-tert-Amylphenol, 99%, Thermo Scientific Chemicals
4-tert-Amylphenol, 99%, Thermo Scientific Chemicals

4-tert-Amylphenol, 99%, Thermo Scientific Chemicals

CAS: 80-46-6 | C11H16O | 164.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number 167330050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS80-46-6
IUPAC Name4-(2-methylbutan-2-yl)phenol
Molecular FormulaC11H16O
InChI KeyNRZWYNLTFLDQQX-UHFFFAOYSA-N
SMILESCCC(C)(C)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Coarse powder or flakes or briquettes
Melting point>=91°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only