3-(3,4-Dihydroxyphenyl)-L-alanine, 99%
3-(3,4-Dihydroxyphenyl)-L-alanine, 99%
3-(3,4-Dihydroxyphenyl)-L-alanine, 99%
Thermo Scientific Chemicals

3-(3,4-Dihydroxyphenyl)-L-alanine, 99%

3-(3,4-Dihydroxyphenyl)-L-alanine (levodopa orL-dopa), CAS # 59-92-7, is a dopamine precursor molecule. | CAS: 59-92-7 | C9H11NO4 | 197.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 167530050
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.10
Save 10.45
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.10
Save 10.45
Each
Chemical Identifiers
CAS59-92-7
IUPAC Name(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO4
InChI KeyWTDRDQBEARUVNC-LURJTMIESA-N
SMILESN[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white to cream
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body

• Upon conversion to dopamine, it acts as an excitatory neurotransmitter

Applications

• L-dopa is useful in the laboratory study of Parkinson’s to replenish dopamine in depleted circuits

• L-dopa has also been used for laboratory research on hypertension and melanocyte activity

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Maini Rekdal, V.; Bess, E.N.; Bisanz, J.E.; Turnbaugh, P.J.; Balskus EP. Discovery and inhibition of an interspecies gut bacterial pathway for Levodopa metabolism. Science. 2019, 364(6445), eaau6323.
  2. Hong, C.T.; Chan, L.; Bai, C.H. The Effect of Caffeine on the Risk and Progression of Parkinson's Disease: A Meta-Analysis. Nutrients. 2020, 12(6), 1860.