Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Thermo Scientific™

Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

The Thermo Scientific™ Nunc™ Edge 2.0 Plate provides an efficient solution for preventing evaporation and in turn the risks associated with edge effect.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
167542160 Microplates
16742550 Microplates
Catalog number 167542
Price (USD)
432.65
Online exclusive
465.00
Save 32.35
Case of 160
Quantity:
160 Microplates
Price (USD)
432.65
Online exclusive
465.00
Save 32.35
Case of 160
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Features
  • The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
  • Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
  • The non-treated surface is available for suspension cell culture or general solution-based assays
  • Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
CertifiedNunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Culture Area0.35 cm2/well
LidWith Lid
No. per Case160
No. per Pack10
Packaging10 Pk., 160 Cases
Product TypeMicroplate
Quantity160 Microplates
Skirt StyleLow Flange
Volume (Metric) Well400 μL
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 160

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .