1,2-Dichloroethane, 99+%, for spectroscopy
1,2-Dichloroethane, 99+%, for spectroscopy
1,2-Dichloroethane, 99+%, for spectroscopy
Acros Organics

1,2-Dichloroethane, 99+%, for spectroscopy

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 167765000
also known as 16776
HSN 29031500
Price (INR)
5,313.00
Each
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
5,313.00
Each
Chemical Identifiers
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
UVat 250 nm A: =<0.15
Residue after evaporation=<0.002 %
UVat 230 nm A: =<1
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only