N,N-Dimethylacetamide, 99+%, for spectroscopy
N,N-Dimethylacetamide, 99+%, for spectroscopy
N,N-Dimethylacetamide, 99+%, for spectroscopy
Thermo Scientific Chemicals

N,N-Dimethylacetamide, 99+%, for spectroscopy

CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 167782500
Price (USD)
67.20
Each
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
67.20
Each
Chemical Identifiers
CAS127-19-5
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.0 %
Water=<0.05 %
UVat 270 nm A: =<1
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only