Ethyl acetate, 99.5+%, for spectroscopy
Ethyl acetate, 99.5+%, for spectroscopy
Ethyl acetate, 99.5+%, for spectroscopy
Thermo Scientific Chemicals

Ethyl acetate, 99.5+%, for spectroscopy

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 167810010
Price (JPY)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.05 %
UVat 255 nm A: =<1
UVat 275 nm A: =<0.05
Appearance (Form)Clear liquid
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only