Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals

CAS: 67-68-5 | Molecular Formula: C2H6OS | Molecular Weight: 78.13 g/mol
Catalog NumberQuantityPackaging
167851000100 mLGlass Bottle
Catalog number 167851000
Price (JPY)
-
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UVat 270 nm A: =<0.5
Water=<0.1 %
UVat 300 nm A: =<0.1
Appearance (Form)Clear liquid
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

RUO – Research Use Only